missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PEGASUS-90 Auto Closing
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3024200230722
Code : NEW
Additional Details : Weight : 170.00000kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested door after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
1072 mm | |
Safety storage cabinet | |
450 mm | |
Powder-coated plain | |
596 mm | |
214 kg | |
Yellow | |
1298 mm | |
S90.129.060.WDAC | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
3 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
524 mm | |
Steel | |
616 x 596 x 1298 mm | |
524 x 450 x 1072 mm | |
616 mm | |
Wing, Left-hinged, Auto-close | |
1 |