missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Type 5/6 Classic Chemical Protection Suit with Feet

Product Code. p-7228476 Shop All Uvex Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17717739 Small Pack of 50
17727739 Medium Pack of 50
17737739 Large Pack of 50
17747739 X-Large Pack of 50
17757739 2X-Large Pack of 50
17767739 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17717739 Supplier Uvex™ Supplier No. 9875909
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Uvex type 5/6 classic disposable suit offers reliable protection with first-class comfort.

  • PP spunbond non-woven fabric with PE film laminated
  • Extremely lightweight and breathable material guarantees an outstanding combination of comfort and safety
  • Soft non-woven material lining
  • Inner overlock seam for outstanding protection against particles and liquid splashes
  • Middle finger loops prevent the sleeve from riding up
  • Optimum fit for a high level of wearer comfort
  • Safe and comfortable hems thanks to elasticated bands in the hood and sleeves
  • Integrated boot sock
  • Particle-tight and limited spray-tightness
  • Anti-static properties
  • Protection from infectious agents
  • Free from paint-wetting impairement substances
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Socks
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance Chemical protection/PPE category III, EN ISO 13982-1:2004+A1:2010 (type 5), EN 13034:2005+A1:2010 (type 6), EN 1149-5:2008, EN 14126:2003+AC:2004, EN 1073-2:2002
For Use With (Application) Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.