missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyk2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£168.00 - £366.00
Specifications
| Antigen | Tyk2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18384674
|
Novus Biologicals
NBP3-16896-20UL |
20 μg |
£168.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18005855
|
Novus Biologicals
NBP3-16896-100UL |
100 μg |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tyk2 Polyclonal antibody specifically detects Tyk2 in Mouse samples. It is validated for Western BlotSpecifications
| Tyk2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Kinase | |
| PBS, 50% glycerol, pH7.3 | |
| 7297 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| EC 2.7.10, EC 2.7.10.2, JTK1, non-receptor tyrosine-protein kinase TYK2, tyrosine kinase 2 | |
| A synthetic phosphorylated peptide around Y1054 & Y1055 of human Tyk2 (NP_003322.3). HEYYRV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title