missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyk2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-16896-100UL
This item is not returnable.
View return policy
Description
Tyk2 Polyclonal antibody specifically detects Tyk2 in Mouse samples. It is validated for Western Blot
Specifications
| Tyk2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| EC 2.7.10, EC 2.7.10.2, JTK1, non-receptor tyrosine-protein kinase TYK2, tyrosine kinase 2 | |
| A synthetic phosphorylated peptide around Y1054 & Y1055 of human Tyk2 (NP_003322.3). HEYYRV | |
| 100 μg | |
| Phospho Specific, Protein Kinase | |
| 7297 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction