missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Tygon™ E-LFL Microbore 2-Stop Tube Sets

Longest life of all Tygon™ formulations

Brand:  Ismatec™ 06447-34

Code : NEW

Additional Details : Weight : 0.04760kg

 View more versions of this product

Product Code. 15491690

  • £289.00 / Pack of 12
Estimated Shipment: 27-09-2024
to see stock.
Add to basket

Description

Description

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • For use with Masterflex™ L/S™ multichannel pump heads
Specifications

Specifications

L/S Series Stopped Pump Tubing
1.42 mm
1.25 ft.
0°C
34
25.3 psi, 1.75 bar
USP; EU Food 10/2011; European Pharmacopoeia; Non-DEHP; Non-Phthalate; RoHS; Prop 65 NA (no Prop 65 chemicals)
Tygon E-LFL
0.38 m
Pk. of 12
67 mL/min
40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers