missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 F Plus with Dissipative Socks Model CHZ5
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 10
Description
- DuPont™ Tychem™ 6000 F Plus with dissipative socks, TFCHZ5TGY18.Hooded coverall. Stitched and over-taped seams. Double cuffs. Knitted inner cuffs. Thumb loops. Elastication at wrists, ankles, face, and waist. Double zippers and double flaps. Outer flap with adhesive tape. Integrated dissipative socks. Grey.
- Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
- Tychem™ 6000 F garment are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Specifications
Specifications
| Product Type | Hooded Coveralls with Socks |
| Size | 4X-Large |
| Gender | Unisex |
| Material | HDPE |
| Color | Gray |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Double |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction