Learn More
DuPont™ Tyvek™ 400 Dual TG Model TD148S
Product Code. 30305964
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30305964
Brand: DuPont™ D15566858
Hooded coverall with Tyvek at the front and large breathable SMS at the back. Stitched and taped seams. Attached socks and undergloves.
  • DuPont™ Tyvek™ 400 Dual TG. Hooded coverall combining Tyvek™ at the front with a SMS nonwoven on the back panel. Available in sizes SM to 3X. Attached Tyvek™ under gloves. Attached Tyvek™ socks. 2-piece hood and gusset for optimal fit. Elasticated hood and waist. White.
  • Tyvek™ front provides an ideal balance of protection, durability and comfort. It is permeable to both air and water vapor, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres down to 1 micron in size on the Tyvek™ front. It is ultra-low-linting and antistatically treated. Silicone non-added. The large, breathable back panel, made of SMS nonwoven, offers lower protection against particles and light water-based splashes, yet high levels of comfort.
  • Tyvek™ 400 Dual TG is designed for specific applications that demand comfort while helping to provide protection from frontal exposure during applications such as mixing/handling of chemicals in chemical industry and for EV battery manufacturing in particular, as well as paint spraying and tasks involving fine hazardous particles or composite materials.
  • Thanks to the taped underarm seams on the upper body, it offers increased protection also for the chemicals/products/process against potential contamination from the wearer or from garment seams during the typical work activity.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size Large
Gender Unisex
Material HDPE
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Socks
Cuff Style Attached Gloves
Neck Style Hooded
Hood Type Elastic
Closure Type Front Zipper with Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Liquid