missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 6000 F Plus
GREENER_CHOICE

Product Code. 15775693 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15775693 Small Pack of 10
15785693 Medium Pack of 10
15795693 Large Pack of 10
15705703 X-Large Pack of 10
15715703 2X-Large Pack of 10
15725703 3X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15775693 Supplier DuPont™ Supplier No. D15344168
  • ONLINE EXCLUSIVE PRICE
    £496.56
    £370.38 / Pack of 10
    Save £126.18
    25% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

DuPont™ Tychem™ 6000 F Plus garments offer excellent chemical permeation resistance to numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals, particulates, biohazards and certain chemical warfare agents, in an innovative design.

  • DuPont™ Tychem™ 6000 F Plus. Hooded coverall available in grey. Elasticated face, wrists, waist and ankles for an optimal fit. Double cuffs, knitted inner cuffs, thumb loops to prevent sleeves from riding up. Front entry with double zipper, double flap and adhesive tape at outer flap.
    Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents. 
    Tychem™ 6000 F garment are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
  • Certified according to tradeulation (EU) 2016/425
  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on inside
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Front closure with double zipper, double flap, outer flap with adhesive tape
  • Double cuffs, thumb loops, knitted inner cuffs
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Tychem
Color Gray
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Double
Neck Style Hooded
Hood Type Elastic
Closure Type Double Zipper with Double Flaps
Seam Type Stitched, Taped
Sterility Non-sterile
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B EN 14126 (barrier to infective agents), EN 1073-2 (protection againstradioactive contamination)
Resistance Chemicals, Liquid, Particle, Anti-static, Abrasion
Collar Type Hooded
Certifications/Compliance Certified according to Regulation (EU) 2016/425, Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), Antistatic treatment (EN 1149-5) - on inside
Description Tychem™
No. of Pockets None
Size (English) Chest 33-36 in.
Size (Metric) Chest 84 to 92 cm
Thickness 210 μm
For Use With (Application) Chemical spill clean-up, emergency response, to military and petrochemical applications.
For Use With (Equipment) Full-face mask
Height (Metric) 162 to 170 cm
Weight (English) 120 g/m2
Quantity 10/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.