missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 S with Socks
GREENER_CHOICE

Product Code. 15959416 Shop All DuPont Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 15
This item is not returnable. View return policy
MTO: Made to order terms & conditions apply. Working in Ex-Zones: Please take this into account for your risk-assessment that the attached socks may isolate the wearer. There is the possibility that the garment and wearer cannot by grounded via the shoes and other measures for grounding the garment and the wearer are required.The garment does not protect against ionizing radiation.This garment and/or fabric are not flame resistant and should not be used around heat, open flame, sparks or in potentially flammable environments.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15959416

Brand: DuPont™ D15193543

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
MTO: Made to order terms & conditions apply. Working in Ex-Zones: Please take this into account for your risk-assessment that the attached socks may isolate the wearer. There is the possibility that the garment and wearer cannot by grounded via the shoes and other measures for grounding the garment and the wearer are required.The garment does not protect against ionizing radiation.This garment and/or fabric are not flame resistant and should not be used around heat, open flame, sparks or in potentially flammable environments.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tychem™ 4000 garments are made from a lightweight, supple and durable fabric. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.

  • DuPont™ Tychem™ 4000 S with socks. Hooded coverall with integrated socks and additional knee-length boot flaps. Available in white, in sizes SM to 3X. Elasticated face, wrists, waist and ankles for an optimal fit, thumb loops to prevent sleeves from riding up. Self-adhesive chin flap for tight seal of suit to face-mask.
    Tychem™ 4000 garments are made from a lightweight and durable fabric (<800g per garment) consisting of a chemical barrier film laminated to a heavy duty Tyvek™ substrate. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.
    Tychem™ 4000 S is ideal for chemical handling, environmental clean-up operations and emergency response. It is suitable for use in a variety of industries, including oil and gas, chemical engineering, and for use by hazardous material response teams and other emergency services.
  • For enhanced liquid protection, taping of outer cuff to glove is recommended. A double cuff is not a substitute for supplementary taping.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size 2X-Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Elastic
Closure Type Double Zipper Front with Double Flaps
Seam Type Stitched, Taped
Sterility Non-sterile
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B
Resistance Chemicals, Liquid, Particle, Anti-static, Abrasion
Certifications/Compliance Certified according to Regulation (EU) 2016/425Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-BEN 14126 (barrier to infective agents), EN 1073-2 (protection againstradioactive contamination)Antistatic treatment (EN 1149-5) - on inside;
Description Tychem 4000 S with socks
No. of Pockets None
Size (English) Chest 46-49 in.
Size (Metric) Chest 116 to 124 cm
For Use With (Application) Chemical handling, environmental clean-up operations and emergency response.
Height (Metric) 186 to 194 cm
Weight (English) 124 g/m2
Quantity 15/Pk.
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.