missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNIP Antibody (JY2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 21 publication
£481.00
Specifications
| Antigen | TXNIP |
|---|---|
| Clone | JY2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Flow Cytometry 2-5 μg/0.1x106 cells, Immunohistochemistry, Immunoprecipitation 2 μg/200ul of cell extract from 5x106 cells, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen reported by customer review, Proximity Ligation Assay, CyTOF-ready, Knockout Validated reported in scientific literature (Brocker et al) |
| Applications | Western Blot |
Description
TXNIP Monoclonal specifically detects TXNIP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, CyTOF-ready, Knockout Validated.Specifications
| TXNIP | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9H3M7 | |
| 10628 | |
| Human recombinant TXNIP | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| JY2 | |
| Western Blot 1 μg/mL, Flow Cytometry 2-5 μg/0.1x106 cells, Immunohistochemistry, Immunoprecipitation 2 μg/200ul of cell extract from 5x106 cells, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen reported by customer review, Proximity Ligation Assay, CyTOF-ready, Knockout Validated reported in scientific literature (Brocker et al) | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with No Preservative | |
| HHCPA78, THIF, thioredoxin binding protein 2, thioredoxin interacting protein, Thioredoxin-binding protein 2, thioredoxin-interacting protein, upregulated by 1,25-dihydroxyvitamin D-3, VDUP1EST01027, Vitamin D3 up-regulated protein 1 | |
| TXNIP | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title