missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNIP Antibody (JY2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 21 publication
Brand: Novus Biologicals NBP1-54578
This item is not returnable.
View return policy
Description
TXNIP Monoclonal specifically detects TXNIP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, CyTOF-ready, Knockout Validated.
Specifications
| TXNIP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Flow Cytometry 2-5 μg/0.1x106 cells, Immunohistochemistry, Immunoprecipitation 2 μg/200ul of cell extract from 5x106 cells, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen reported by customer review, Proximity Ligation Assay, CyTOF-ready, Knockout Validated reported in scientific literature (Brocker et al) | |
| Q9H3M7 | |
| TXNIP | |
| Human recombinant TXNIP | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| JY2 | |
| Unconjugated | |
| PBS with No Preservative | |
| HHCPA78, THIF, thioredoxin binding protein 2, thioredoxin interacting protein, Thioredoxin-binding protein 2, thioredoxin-interacting protein, upregulated by 1,25-dihydroxyvitamin D-3, VDUP1EST01027, Vitamin D3 up-regulated protein 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10628 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction