Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased
GREENER_CHOICEBIOPHARMA
Product Code. 18078794 Shop All Eppendorf Products
Click to view available options
Color:
Clear
Colorless
Green
Skirt Style:
Semi-skirted
Skirted
Product Type:
BioBased LoBind PCR Plates
BioBased PCR Plates
No. of Wells:
96
384
Unit Size:
Pack of 25
Product Code. 18078794
Brand: Eppendorf™ 0030531060
Only null left
Add to basket
Add to basket
Discover the new twin.tec™ PCR Plates BioBased made of bio-based polymer (mass balance approach). These plates allow you to significantly reduce your consumable-related carbon footprint while maintaining identical technical performance.
  • Reduction of consumable-related carbon footprint in the lab
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
  • Available with barcode (upon request)
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Array 8 x 12
Binding Type LoBind
Product Line Eppendorf
Color Colorless
Product Type BioBased LoBind PCR Plates
Skirt Style Semi-skirted
Material Polypropylene, Polycarbonate
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Quantity 25 Pk.
Fit for Purpose PCR Ready