Learn More
Honeywell™ Turboshield Hard Hat Adapter
Product Code. 15856665
Change view
Click to view available options
:
For use with hard hats, Turboshield face shield
Product Code.
15856665 For use with hard hats, Turboshield face shield
1 options
Product Code. 15856665 Supplier Honeywell™ Supplier No. 1031749
Only null left
Add to basket
Add to basket
Universal hard hat adapter for Turboshield face protection system with patented dual-pivot mechanism and visor latch release.
  • Features a patented dual-pivot mechanism and a patented visor latch mechanism release system
  • Perfect for workers exposed to falling for flying objects, impact, splash, UV and airborne debris
  • Hard hat adapter option makes it easier for those who need to combine head protection with face protection
  • Visors slide back 170 mm, improving balance and weight distribution
  • Dual-hinge design increases clearance by as much as 100 mm and keeps accessory slots open for compatibility with other PPE
  • Compatible with Supervisor specialty visors including gold-coated, triacetate and acetate
  • Compatible with most major hard hats;(including North brand) with the exception of full brim hard hats.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 2, EC Certificate: C1519.2HON, EC Attestation: C1519.2HON, EN 166
Description Dielectric
For Use With (Equipment) Hard hats, Turboshield face shield
For Use With (Application) Impact protection, splash protection, UV protection
Frame Color Gray
Frame Material Reinforced Nylon