missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Turbo3C Protease

Product Code. 16191840
Click to view available options
:
1 mg
This item is not returnable. View return policy

Product Code. 16191840

Brand: Abnova™ P5380.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human rhinovirus (HRV) 3C protease with GST and His tags expressed in Escherichia coli.

Specifications

Concentration 2 mg/mL
For Use With (Application) Enzyme
Content And Storage Store at -20°C, Aliquot to avoid repeated freezing and thawing
Form Liquid
Quantity 1 mg
Product Type Protein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.