Learn More
RETSCH Tungsten Carbide Grinding Jars
Product Code. 10212772 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
For EMAX
For MM 200 Mixer Mill
For MM 301 Mixer mill
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
Unit Size:
Each
Product Code. 10212772
Brand: RETSCH 01.462.0235
Only null left
Add to basket
Add to basket
Use these grinding jars for dry, wet and cryogenic grinding of small samples. Retsch™ Tungsten Carbide Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
  • Screw-top Tungsten grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Product Type Grinding Jar
Capacity (Metric) 10 mL
For Use With (Equipment) For MM 301 Mixer mill
Material Tungsten Carbide