missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Tungsten Carbide Grinding Balls

Product Code. 10416811 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
MM 200 and MM 301
MM 200 and MM 400
Planetary Ball Mill PM 400
Reaction Vials
Diameter (Metric):
3 mm
4 mm
5 mm
7 mm
10 mm
12 mm
15 mm
20 mm
30 mm
40 mm
Unit Size:
Each
Pack of 200
This item is not returnable. View return policy

Product Code. 10416811

Brand: RETSCH 05.368.0071

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Grinding Balls
Diameter (Metric) 10 mm
For Use With (Equipment) Planetary Ball Mill PM 400
Material Tungsten Carbide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.