Learn More
RETSCH Tungsten Carbide Comfort Grinding Jars
Use these grinding jars with Retsch™ ball mills. Retsch™ Tungsten Carbide Comfort Grinding Jars feature an O-type sealing for safe operation.
Brand: RETSCH 01.462.0222
Description
- Available in a variety of sizes
- Optional aeration covers
- Pressure tight safety closure devices
Specifications
Comfort Grinding Jar | |
250 mL | |
For PM 100, PM 400 | |
Tungsten Carbide |