Learn More
R&D Systems™ TumorTACS In Situ Apoptosis Detection Kit
Product Code. 18411320
Change view
Click to view available options
Quantity:
30 Tests
Unit Size:
30 tests
Product Code. Quantity unitSize
18411320 30 Tests 30 tests
1 options
Product Code. 18411320 Supplier R&D Systems™ Supplier No. 481530K
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Streptavidin-HRP, DAB, Methyl Green counterstain, and reagents necessary for generating positive controls
Designed for detection of DNA fragmentation in fixed cells, embedded tissue, and frozen tissue using light microscopy
A common biochemical marker of apoptosis is DNA fragmentation. In situ, the 3 ft. ends of cleaved DNA fragments provide a substrate for the enzyme terminal deoxynucleotidyl transferase (TdT) which adds nucleotides at the site of DNA breaks.
Optimized for detection of apoptosis in tumor-derived sections and cells
Supplemented with a cation that specifically enhances the signal in apoptotic tumor cells
TACS-Nuclease™ allows positive controls to be
generated for each experim
TRUSTED_SUSTAINABILITY
Specifications
Quantity 30 Tests
Content And Storage The product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Do not use past expiration date.