missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tubulin Tracker™ Deep Red
Tubulin Tracker™ Deep Red provides deep-red/far-red fluorescence when bound to polymerized tubulin in live cells.
£141.00 - £449.00
Specifications
Excitation Wavelength Range | 652⁄669 |
---|---|
Product Line | TubulinTracker™ |
Shipping Condition | Approved for shipment on Wet or Dry Ice |
Label Type | UV-Vis Labels |
Product Type | Tubulin Tracker |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16190561
|
Invitrogen™
T34077 |
60 slides |
£141.00
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16180561
|
Invitrogen™
T34076 |
300 slides |
£449.00
1 set |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Based on Docetaxel conjugated with a bright, photostable deep-red fluorophore
- Docetaxel belongs to the family of cytoskeletal drugs that target tubulin
- Absorbs and emits optimally at 652 nm and 669 nm, respectively and can be visualized with standard Cy5 filter settings using almost any fluorescent imaging instrument
- Can be multiplexed with blue, green, orange, red, and near-IR fluorophores.
Specifications
652⁄669 | |
Approved for shipment on Wet or Dry Ice | |
Tubulin Tracker |
TubulinTracker™ | |
UV-Vis Labels | |
Tubulin |
For Research Use Only. Not for use in diagnostic procedures.