missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TUBA1B/TUBA4A Recombinant Rabbit Monoclonal Antibody (24GB2020)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen TUBA1B/TUBA4A
Clone 24GB2020
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274793
GREENER_CHOICE
View Documents
Invitrogen™
MA557299
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275288
View Documents
Invitrogen™
MA557300
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TUBA1B/TUBA4A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

TUBA1B and TUBA4A are genes that encode for alpha-tubulin isoforms, essential components of the microtubule system that plays a critical role in maintaining cell structure, intracellular transport, and cell division. TUBA1B, also known as tubulin alpha-1b chain, is located on chromosome 12q13.12 and has been linked to various cancers due to its influence on microtubule dynamics and cellular proliferation. It is also associated with immune cell infiltration and modulation of immune checkpoints, making it significant in tumor growth and metastasis. TUBA4A, known as tubulin alpha-4A, is found on chromosome 2q35 and is linked to neurodevelopmental disorders and neurodegenerative diseases such as amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD). Mutations in TUBA4A can lead to defects in microtubule stability and function, emphasizing its role in neuronal health. Both genes highlight the critical role of microtubules in cellular and neurological functions, and their mutations can contribute to diverse pathological conditions, including cancer and neurodegenerative diseases.
Specifications

Specifications

TUBA1B/TUBA4A
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
TUBA1B
10376, 22143, 22145, 316531, 500929, 7277
A synthesized peptide derived from human TUBA1B/4A (aa 33-47).
Antibody
24GB2020
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P05213, P68363, P68366, P68368, Q5XIF6, Q6P9V9
TUBA1B, TUBA4A
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.