missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTP 22, Tocris Bioscience™
High affinity, selective CK2 inhibitor
£192.56 - £730.40
Chemical Identifiers
CAS | 329907-28-0 |
---|---|
Molecular Formula | C16H14N2O2S2 |
Molecular Weight (g/mol) | 330.42 |
MDL Number | MFCD02654497 |
InChI Key | RAOULLCLLOGTDA-UHFFFAOYSA-N |
PubChem CID | 1536915 |
IUPAC Name | 3-{[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanyl}propanoic acid |
SMILES | CC1=CC=C(C=C1)C1=CSC2=NC=NC(SCCC(O)=O)=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18004142
|
Tocris Bioscience
4432/10 |
10 mg |
£192.56
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18075532
|
Tocris Bioscience
4432/50 |
50 mg |
£730.40
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
High affinity, ATP-competitive casein kinase 2 (CK2) inhibitor (IC50 = 0.1 μM, Ki = 40 nM). Displays selectivity for CK2 over JNK3, ROCK1 and MET (no inhibitory effects at 10 μM).Chemical Identifiers
329907-28-0 | |
330.42 | |
RAOULLCLLOGTDA-UHFFFAOYSA-N | |
3-{[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanyl}propanoic acid |
C16H14N2O2S2 | |
MFCD02654497 | |
1536915 | |
CC1=CC=C(C=C1)C1=CSC2=NC=NC(SCCC(O)=O)=C12 |
Specifications
329907-28-0 | |
MFCD02654497 | |
RAOULLCLLOGTDA-UHFFFAOYSA-N | |
3-{[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanyl}propanoic acid | |
1536915 | |
>98% |
C16H14N2O2S2 | |
ttp 22, ttp22, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl sulfanylpropanoic acid, ttp-22, 3-5-p-tolyl thieno 2,3-d pyrimidin-4-yl thio propanoic acid, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl sulfanyl propanoic acid, 7ey, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl thio propanoic acid, 3-5-4-methylphenyl thiopheno 3,2-e pyrimidin-4-ylthio propanoic acid | |
CC1=CC=C(C=C1)C1=CSC2=NC=NC(SCCC(O)=O)=C12 | |
330.42 | |
330.42 | |
TTP 22 |
Safety and Handling
Recommended Storage : Store at +4°C