missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TTP 22

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
High affinity, ATP-competitive casein kinase 2 (CK2) inhibitor (IC50 = 0.1 μM, Ki = 40 nM). Displays selectivity for CK2 over JNK3, ROCK1 and MET (no inhibitory effects at 10 μM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 330.42 |
| Purity | >98% |
| Molecular Weight (g/mol) | 330.42 |
| Chemical Name or Material | TTP 22 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction