CAS RN 329907-28-0
CAS RN 329907-28-0
IUPAC Name:
3-{[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanyl}propanoic acid
Synonyms:
3-{[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanyl}propanoic acid
Molecular Weight (g/mol):
330.42
Molecular Formula:
C16H14N2O2S2
InChi Key:
RAOULLCLLOGTDA-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=CSC2=NC=NC(SCCC(O)=O)=C12
1
–
2
of
2
results
1
Tocris Bioscience™ TTP 22
CAS: 329907-28-0 Molecular Formula: C16H14N2O2S2
2
TTP 22, MedChemExpress
MedChemExpress TTP 22 is a potent CK2 inhibitor, with an IC50 of 100 nM and a Ki of 40 nM.