missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC9C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55306
This item is not returnable.
View return policy
Description
TTC9C Polyclonal specifically detects TTC9C in Human samples. It is validated for Western Blot.
Specifications
| TTC9C | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MGC29649, tetratricopeptide repeat domain 9C, tetratricopeptide repeat protein 9C, TPR repeat protein 9C | |
| Rabbit | |
| 20 kDa | |
| 100 μL | |
| Primary | |
| Pig: 86%; Guinea pig: 77%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8N5M4 | |
| TTC9C | |
| Synthetic peptides corresponding to TTC9C(tetratricopeptide repeat domain 9C) The peptide sequence was selected from the N terminal of TTC9C. Peptide sequence MEKRLQEAQLYKEEGNQRYREGKYRDAVSRYHRALLQLRGLDPSLPSPLP. | |
| Affinity purified | |
| RUO | |
| 283237 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction