missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55108
This item is not returnable.
View return policy
Description
TTC8 Polyclonal specifically detects TTC8 in Human samples. It is validated for Western Blot.
Specifications
| TTC8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Bardet-Biedl syndrome 8 protein, BBS8Bardet-Biedl syndrome type 8, RP51, tetratricopeptide repeat domain 8, tetratricopeptide repeat protein 8, TPR repeat protein 8 | |
| Rabbit | |
| 54 kDa | |
| 100 μL | |
| Cancer, Vision | |
| 123016 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8TAM2-3 | |
| TTC8 | |
| Synthetic peptides corresponding to TTC8(tetratricopeptide repeat domain 8) The peptide sequence was selected from the N terminal of TTC8. Peptide sequence ENAIAQVPRPGTSLKLPGTNQTGGPSQAVRPITQAGRPITGFLRPSTQSG. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction