missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSH beta Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £389.00
Specifications
| Antigen | TSH beta |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17909382
|
Invitrogen™
PA5145294 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17978772
![]() |
Invitrogen™
PA5145293 |
200 μL |
£389.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.Specifications
| TSH beta | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Canine | |
| P54828 | |
| 403973 | |
| Recombinant Dog TSHb protein, Ser20-Ile138 (Accession #P54828). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| thyroid stimulating hormone beta; thyroid stimulating hormone subunit beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone beta subunit; thyroid-stimulating hormone subunit beta; Thyrotropin alfa; Thyrotropin beta chain; thyrotropin beta subunit; thyrotropin subunit beta; TSH; Tshb; TSH-B; TSH-beta | |
| TSHB | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| TSHB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title