missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSH beta Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145293
This item is not returnable.
View return policy
Description
TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.
Specifications
| TSH beta | |
| Polyclonal | |
| Unconjugated | |
| TSHB | |
| thyroid stimulating hormone beta; thyroid stimulating hormone subunit beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone beta subunit; thyroid-stimulating hormone subunit beta; Thyrotropin alfa; Thyrotropin beta chain; thyrotropin beta subunit; thyrotropin subunit beta; TSH; Tshb; TSH-B; TSH-beta | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 403973 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P54828 | |
| TSHB | |
| Recombinant Dog TSHb protein, Ser20-Ile138 (Accession #P54828). | |
| 200 μL | |
| Primary | |
| Canine | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu