missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSFM Recombinant Rabbit Monoclonal Antibody (23GB4710), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | TSFM |
|---|---|
| Clone | 23GB4710 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088288
![]() |
Invitrogen™
MA553792 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089261
|
Invitrogen™
MA553793 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSFM Recombinant Monoclonal antibody specifically detects TSFM in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| TSFM | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P43897, Q9CZR8, Q9QYU2 | |
| 10102, 66399, 679068 | |
| IgG | |
| Affinity Chromatography | |
| Antibody |
| 23GB4710 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2310050B20Rik; 2A3-2; 9430024O13Rik; EFTS; EF-Ts; EF-Ts {ECO:0000255; EFTSMT; EF-TsMt; EF-TsMt {ECO:0000255; elongation factor Ts; elongation factor Ts, mitochondrial; elongation factor Ts, mitochondrial {ECO:0000255; HAMAP-Rule:MF_03135}; mitochondrial elongation factor Ts; translation elongation factor; Ts translation elongation factor, mitochondrial; TSFM; zgc:158429 | |
| A synthesized peptide derived from human TSFM (40-66AA). | |
| Primary | |
| -20°C | |
| TSFM |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title