missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSFM Recombinant Rabbit Monoclonal Antibody (23GB4710), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553793
This item is not returnable.
View return policy
Description
Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 ).
Specifications
| TSFM | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TSFM | |
| 2310050B20Rik; 2A3-2; 9430024O13Rik; EFTS; EF-Ts; EF-Ts {ECO:0000255; EFTSMT; EF-TsMt; EF-TsMt {ECO:0000255; elongation factor Ts; elongation factor Ts, mitochondrial; elongation factor Ts, mitochondrial {ECO:0000255; HAMAP-Rule:MF_03135}; mitochondrial elongation factor Ts; translation elongation factor; Ts translation elongation factor, mitochondrial; TSFM; zgc:158429 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10102, 66399, 679068 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4710 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43897, Q9CZR8, Q9QYU2 | |
| TSFM | |
| A synthesized peptide derived from human TSFM (40-66AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction