missing translation for 'onlineSavingsMsg'
Learn More

TSC22D2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen TSC22D2
Concentration 0.39 mg/mL
Applications Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16843995
View Documents
Proteintech
25418-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16833995
View Documents
Proteintech
25418-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TSC22D2 belongs to the TSC-22/Dip/Bun family and the function remains unknown.
Specifications

Specifications

TSC22D2
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Unconjugated
Rabbit
Human
O75157
9819
TSC22D2 Fusion Protein Ag22294
Primary
-20°C
TSC22D2
0.39 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
KIAA0669, TILZ4, TILZ4a, TILZ4b, TILZ4c, TSC22 domain family protein 2, TSC22 domain family, member 2, TSC22D2
TSC22D2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.