missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC22D2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25418-1-AP-150UL
This item is not returnable.
View return policy
Description
TSC22D2 belongs to the TSC-22/Dip/Bun family and the function remains unknown.Specifications
| TSC22D2 | |
| Polyclonal | |
| Unconjugated | |
| TSC22D2 | |
| KIAA0669, TILZ4, TILZ4a, TILZ4b, TILZ4c, TSC22 domain family protein 2, TSC22 domain family, member 2, TSC22D2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9819 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75157 | |
| TSC22D2 | |
| TSC22D2 Fusion Protein Ag22294 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction