missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSC Agar Base

Product Code. 13235819
Change view
Click to view available options
Fill Weight:
19.5 to 20.5 g
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight unitSize
13235819 19.5 to 20.5 g Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13235819 Supplier Thermo Scientific™ Supplier No. TV5204G
  • ONLINE EXCLUSIVE PRICE
    £124.00
    £120.64 / Pack of 50
    Save £3.36
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selectively isolate and enumerate Clostridium perfringens in food and water samples by detecting sulfite reduction, which produces black colonies under anaerobic conditions.

TSC Agar Base is used to selectively isolate and enumerate Clostridium perfringens in food and water samples by detecting sulfite reduction, which produces black colonies under anaerobic conditions.

TRUSTED_SUSTAINABILITY

Specifications

Color Green Beige, Transparent
Content And Storage 2°C to 25°C
Format Poured Tube
Manufacturing Origin Germany
Product Type Agar
Fill Weight 19.5 to 20.5 g
Packaging Type 50 Tubes/Box
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.