missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSB Double Bag (100 mL Septum)

Product Code. 17759448
Change view
Click to view available options
:
100 mL Septum
Product Code.
17759448 100 mL Septum
1 options
This item is not returnable. View return policy

Product Code. 17759448

Brand: Thermo Scientific™ R112986

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Tryptic Soy Broth is general purpose enrichment broth for the cultivation and isolation of microorganisms.

  • Borosilicate bottle
  • Phenolic screw cap with septum
TRUSTED_SUSTAINABILITY

Specifications

Color Amber
Media Type Culture Media
Fill Volume 100 mL
For Use With (Application) Sterility testing
Packaging Type Double bagged
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.