missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Tryptone Soya Broth (Bottled)

Product Code. 15622129
Change view
Click to view available options
Fill Weight:
10.01 to 0.3 g
10.0g + 0.3g
100 to 101.5 g
100.0 - 101.5g
100.0g + 1.5 g
1000 to 1005 g
20.0 - 20.3 g
450 to 454 g
500.0 - 504.0 g
90 to 90.5 g
Unit Size:
1 set
Pack of 10
Pack of 24
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight unitSize
15622129 100 to 101.5 g Pack of 10
12786959 10.0g + 0.3g Pack of 24
15632129 450 to 454 g Pack of 10
12693428 100.0g + 1.5 g Pack of 10
11983112 10.01 to 0.3 g Pack of 24
15239047 20.0 - 20.3 g Pack of 24
12864132 90 to 90.5 g Pack of 10
12854132 100.0 - 101.5g Pack of 10
15832357 1000 to 1005 g 1 set
13255489 500.0 - 504.0 g Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 15622129 Supplier Thermo Scientific™ Supplier No. BO0282M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Grow a wide variety of microprganisms using this medium in convenient ready prepared bottles.

Grow a wide variety of microorganisms with Thermo Scientific™ Tryptone Soya Broth ready-prepared bottled medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Tryptone Soya Broth.

  • Conforms to formulations detailed in International Pharmacopoeia.
  • Convenient: pre-filled bottles are ready for immediate use.
  • Simple: easy to use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 25°C
Format 100 mL DIN bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 100 to 101.5 g
Packaging Type 10 DIN bottles with injectable closures in a box.
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.