missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soya Agar + TWEEN / lecithin55 Contact Plate

Product Code. 12907168
This item is not returnable. View return policy

Product Code. 12907168

Brand: Thermo Scientific™ PO0479D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless surface sampling in the most regulated environments using this Triple Wrapped Irradiated Contact Plate.

Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Agar with TWEEN/Lecithin 55 Irradiated Triple Wrap Contact Plate. The general purpose medium, containing two peptones, supports the growth of wide variety of organisms. It is suitable for the cultivation of both aerobes and anaerobes. Due to the presence of TWEEN 80 and lecithin, the medium can be used as Microbial Test Agar for testing quaternary ammonium compounds1. The medium conforms to the formulations detailed in international pharmacopoeia.

Streamline environmental monitoring of cleanroom environments with Tryptone Soya Agar with TWEEN/Lecithin 55 Irradiated Triple Wrap Contact Plate.

  • Secure: dome-shaped bottom prevents agar from falling out of the dish
  • Easy-to-handle: GripFit™ technology ensures the lid remains in place during handling while being easily removed during environmental monitoring
  • Straightforward results interpretation: inner layer on the plate base is sub-divided into grids to allow easy calculation of colony forming units (CFU) from surface tests, enabling labs to report whether contaminant levels fall within pre-defined acceptable limits
  • Easy-to-stack: provides more stability during handling and transport, and ventilation is reduced to prevent plates from drying out
  • Consistent: manufactured using a specialized, automated procedure to ensure a smooth, evenly distributed layer of agar on each plate
  • Contamination risk reduction: Protective layers enhance moisture control by incorporating novel elements that permit the passage and absorption of excess moisture, reducing the risk of contamination

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, clear
Content And Storage 2 to 25°C
Description Tryptone Soya Agar + TWEEN / lecithin55 Contact Plate
Format 55mm Contact Plate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 13.5g ± 5 %
Packaging Type Triple Wrapped - Irradiated
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.