Learn More
Thermo Scientific™ Tryptone Soya Agar Irradiated (Triple Wrap) Plate
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Oxoid™ Tryptone Soya Agar Irradiated Plate. The Tryptone Soya Agar supports the growth of wide variety of organisms.
Perform effortless environmental monitoring with Tryptone Soya Agar Irradiated (Triple Wrap) Plate.
- Sterile: plates are irradiated and triple wrapped in a transparent packaging.
- Consistent: automated manufacturing procedures ensure a smooth, evenly distributed layer of agar on each plate.
- Optimized: products undergo quantitative quality control testing, using a low dose inoculum to ensure optimum performance.
- Ready-to-use: convinince of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Ivory, transparent |
| Content And Storage | 2 to 25°C |
| Description | Tryptone Soya Agar Irradiated (Triple Wrap) Plate |
| Format | 90mm Plate |
| Manufacturing Origin | Germany |
| Product Type | Agar |
| Fill Weight | 25 g ± 0.5 g |
| Packaging Type | Triple Wrapped - Irradiated |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.