missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soya Agar Contact Plate

Product Code. 11993752
This item is not returnable. View return policy

Product Code. 11993752

Brand: Thermo Scientific™ PO0193C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Grow a wide variety of microorganisms with this formulation in easy-to-handle contact plate format for effortless sample surfacing.

Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Agar Contact Plate. The general purpose medium, containing two peptones, supports the growth of wide variety of organisms. It is suitable for the cultivation of both aerobes and anaerobes. The medium conforms to the formulations detailed in international pharmacopoeia1,2,3,4

Perform effortless surface sampling with the easy-to-handle Tryptone Soya Agar Contact Plate.

  • Easy-to-handle: dome shaped bottom format for effortless surface sampling.
  • Consistent performance: smooth, even agar surface ensured through specialized manufacturing process.
  • Easy-to-stack providing more stability during handling and transport.
  • Conforms to formulations detailed in International Pharmacopoeia.
  • Ready-to-use convenience of prepared media.

Prevent agar from falling out of the dish with Thermo Scientific Contact Plates, which feature a dome-shaped bottom. GripFit™ technology makes plates easy to handle and ensures the lid remains in place during handling, while being easily removed during environmental monitoring. An inner layer on the base of the plate is sub-divided into grids to allow easy calculation of colony forming units (CFU) from surface tests, enabling labs to report whether contaminant levels fall within pre-defined acceptable limits. Easy stacking provides more stability during handling and transport, and ventilation is reduced to prevent plates from drying out.

Our contact plates are manufactured using a specialized, automated procedure to ensure a smooth, evenly distributed layer of agar on each plate. Surface sampling is achieved by gently rolling the domed surface of the agar onto the area to be tested. If any microorganisms that can cause disease or infection are present, they will grow to form easily visible colonies on the surface of the agar. This offers a reliable and effective way of determining the microbial cleanliness of surface areas and help control invisible dangers associated with microbial contamination. Clearly visible growth is available within three days for bacteria and within five days for fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, Clear
Content And Storage 2°C to 12°C
Description Tryptone Soya Agar Contact Plates
Format 55mm Contact Plate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 13.5g ± 5 %
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.