missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soya Agar (Casein Soya Bean Digest Agar)

Product Code. 10649824
Change view
Click to view available options
Media Type:
Tryptone Soya Agar
Fill Volume:
17 mL
19 mL
25 mL
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Media Type Fill Volume Unit Size
10649824 Tryptone Soya Agar 17 mL Pack of 10
10758123 Tryptone Soya Agar 25 mL Pack of 10
12932588 Tryptone Soya Agar 19 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10649824 Supplier Thermo Scientific™ Supplier No. PO5012A
  • ONLINE EXCLUSIVE PRICE
    £10.25
    £9.05 / Pack of 10
    Save £1.20
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grow a wide variety of microorganisms with general purpose Thermo Scientific™ Tryptone Soya Agar prepared media plates.

Grow a wide variety of microorganisms with Thermo Scientific ™ Tryptone Soya Agar. A general purpose agar medium, containing two peptones, Tryptone Soya Agar will support the growth of both aerobes and anaerobes, the latter being grown either in deep cultures or by incubation under anaerobic conditions. Tryptone Soya Agar is recommended as a reference medium when testing selective media to measure the degree of inhibition.

Tryptone Soya Agar, also known as Casein Soya Bean Digest Agar, is a general purpose medium specified in pharmacopoeia and food testing methods for the growth of a wide variety of organisms. The product is tested to the pharmacopoeia and is mainly used as reference medium when testing selective media.
  • Has the ready-to-use convenience of prepared media
  • May be used as a blood agar base
  • Recommended as a reference medium for testing selective media
  • Conforms to formulations detailed in International Pharmacopoeia
Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 12°C, in the dark
Format 90mm Monoplate
Media Type Tryptone Soya Agar
Product Type Agar Plate
Fill Volume 17 mL
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.