missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tryptone Soya Agar
Description
Tryptone Soya Agar slopes may be used to cultivate, store, and ship bacterial cultures.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2 to 25°C |
| Format | 125mL sirop bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 99-101g |
| Packaging Type | Ten sirop bottles with screw cap closures in a box. |
| Quantity | 10/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?