missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Bile X-Glucuronide (TBX) Medium

Product Code. 11974162
Change view
Click to view available options
Quantity:
10/Pk.
Fill Weight:
17g ± 5%
19.5g ± 1.0.g
Media Type:
TBX Medium
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Fill Weight Media Type Unit Size
11974162 10/Pk. 17g ± 5% TBX Medium Pack of 10
12972628 10/Pk. 19.5g ± 1.0.g TBX Medium Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11974162 Supplier Thermo Scientific™ Supplier No. PO5109A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect and enumerate Escherichia coli in food and animal feed samples with this selective, chromogenic, ready-to-use medium.

Detect and enumerate Escherichia coli in food samples with Thermo Scientific™ Typtone Bile X-Glucuronide (TBX) Medium. TBX Medium builds on the speed, reliability and recovery from frozen samples and detection of poor lactose fermenters offered by the original Tryptone Bile Agar1,2, with the addition of a chromogenic agent, X-glucuronide, which detects glucuronidase activity: an enzyme shown to be highly specific for Escherichia coli3. The released chromophore in TBX Medium is insoluble and accumulates within the cell, making colored target colonies easy to identify

Most Escherichia coli strains can be differentiated from other coliforms by the presence of the enzyme glucuronidase. However, approximately 3 to 4% of Escherichia coli are glucuronidase negative, notably Escherichia coli O157 strains4.

The chromogen in TBX Medium is 5-bromo-4-chloro-3-indolyl-beta-D-glucuronide (X-glucuronide) and is targeted by glucuronidase. Escherichia coli cells are able to absorb this complex intact and intracellular glucuronidase splits the bond between the chromophore and the glucuronide. The released chromophore is colored and builds up within the cells, causing Escherichia coli colonies to be colored blue/green.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Ivory, transparent
Content And Storage 2° to 12°C, in the dark
Format 90mm Monoplate
Manufacturing Origin Germany
Media Type TBX Medium
Product Type Agar Plate
Fill Volume 17 mL
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.