Learn More
Abcam Trypsin Activity Assay Kit (Fluorometric)
Product Code. 32120013 Shop All Abcam Products
Change view
Click to view available options
100 Tests
Product Code.
32120013 100 Tests
1 options
Product Code. 32120013 Supplier Abcam Supplier No. ab282916100Test
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure TRYP activity in Cell/Tissue Extracts, Biofluids with Trypsin Activity Assay Kit AB282916. Sensitivity = 2 μU.
Measure TRYP activity in Cell/Tissue Extracts, Biofluids with Trypsin Activity Assay Kit AB282916. Sensitivity = 2 μU.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests