missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypsin 3/PRSS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | Trypsin 3/PRSS3 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697628
|
Novus Biologicals
NBP2-99895-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634277
|
Novus Biologicals
NBP2-99895-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Trypsin 3/PRSS3 Polyclonal antibody specifically detects Trypsin 3/PRSS3 in Human samples. It is validated for ELISASpecifications
| Trypsin 3/PRSS3 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Proteases & Other Enzymes | |
| 0.2 um filtered solution in PBS | |
| 5646 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsinogen, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin III, Trypsin IV, trypsin-3, trypsinogen 4, trypsinogen 5, trypsinogen IV | |
| Produced in rabbits immunized with purified, recombinant Human Trypsin 3/PRSS3 (Accession#: P35030-3; Met 1-Ser 247) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title