missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypsin 3/PRSS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99895-50ul
This item is not returnable.
View return policy
Description
Trypsin 3/PRSS3 Polyclonal antibody specifically detects Trypsin 3/PRSS3 in Human samples. It is validated for ELISA
Specifications
| Trypsin 3/PRSS3 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsinogen, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin III, Trypsin IV, trypsin-3, trypsinogen 4, trypsinogen 5, trypsinogen IV | |
| Produced in rabbits immunized with purified, recombinant Human Trypsin 3/PRSS3 (Accession#: P35030-3; Met 1-Ser 247) | |
| 50 μg | |
| Cancer, Proteases & Other Enzymes | |
| 5646 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction