missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£168.00 - £366.00
Specifications
| Antigen | TRPM1 |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18364824
|
Novus Biologicals
NBP3-04694-20UL |
20 μL |
£168.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18303397
|
Novus Biologicals
NBP3-04694-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPM1 Polyclonal specifically detects TRPM1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TRPM1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 4308 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| CSNB1C, Long transient receptor potential channel 1, LTrpC1, LTRPC1transient receptor potential cation channel subfamily M member 1, melastatin 1, melastatin-1, MLSN, MLSN1transient receptor potential melastatin family, transient receptor potential cation channel, subfamily M, member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1343-1642 of human TRPM1 (NP_001238949.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title