missing translation for 'onlineSavingsMsg'
Learn More

TRPM1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18364824 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364824 20 μL 20µL
18303397 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18364824 Supplier Novus Biologicals Supplier No. NBP30469420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRPM1 Polyclonal specifically detects TRPM1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPM1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CSNB1C, Long transient receptor potential channel 1, LTrpC1, LTRPC1transient receptor potential cation channel subfamily M member 1, melastatin 1, melastatin-1, MLSN, MLSN1transient receptor potential melastatin family, transient receptor potential cation channel, subfamily M, member 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1343-1642 of human TRPM1 (NP_001238949.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4308
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.