missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT61B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | TRMT61B |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
TRMT61B Polyclonal specifically detects TRMT61B in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| TRMT61B | |
| Unconjugated | |
| RUO | |
| DKFZp564I2178, EC 2.1.1.36, FLJ20628, potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B, tRNA methyltransferase 61 homolog B (S. cerevisiae) | |
| TRMT61B | |
| IgG | |
| This product is specific to Subunit or Isoform: TRMT61B. |
| Polyclonal | |
| Rabbit | |
| Human | |
| 55006 | |
| Synthetic peptides corresponding to FLJ20628(hypothetical protein FLJ20628) The peptide sequence was selected from the N terminal of FLJ20628. Peptide sequence PLSISDIGTGCLSSLENLRLPTLREESSPRELEDSSGDQGRCGPTHQGSE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title