missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT61B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-53080
This item is not returnable.
View return policy
Description
TRMT61B Polyclonal specifically detects TRMT61B in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TRMT61B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| TRMT61B | |
| Synthetic peptides corresponding to FLJ20628(hypothetical protein FLJ20628) The peptide sequence was selected from the N terminal of FLJ20628. Peptide sequence PLSISDIGTGCLSSLENLRLPTLREESSPRELEDSSGDQGRCGPTHQGSE. | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: TRMT61B. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunoprecipitation | |
| DKFZp564I2178, EC 2.1.1.36, FLJ20628, potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B, tRNA methyltransferase 61 homolog B (S. cerevisiae) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55006 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction