missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkB, p Tyr707, p Tyr706 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£163.00 - £549.00
Specifications
| Concentration | 1 mg/mL |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Target Species | Human, Mouse, Rat |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18259504
|
Novus Biologicals
NBP2-54764-0.05MG |
0.05 mg |
£345.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18085024
|
Novus Biologicals
NBP2-54764 |
0.1 mg |
£549.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693967
|
Novus Biologicals
NBP2-54764-0.025mg |
0.025 mg |
£163.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TrkB Polyclonal specifically detects TrkB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| 1 mg/mL | |
| Unconjugated | |
| Human, Mouse, Rat | |
| 4915 | |
| Trk B (phospho Tyr706/Tyr707) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 706/tyrosine 707 (T-D-Y (p)-Y (p)-R-V)derived from Human Trk B. | |
| Primary |
| Polyclonal | |
| Rabbit | |
| BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
| NTRK2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title