missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkB, p Tyr707, p Tyr706 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54764
This item is not returnable.
View return policy
Description
TrkB p Tyr706 Polyclonal antibody specifically detects TrkB p Tyr706 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TrkB (p Tyr706) | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| NTRK2 | |
| Trk B (phospho Tyr706/Tyr707) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 706/tyrosine 707 (T-D-Y (p)-Y (p)-R-V)derived from Human Trk B. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody detects endogenous levels of TrkB only when phosphorylated at tyrosine 706 and tyrosine 707. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry 1:10 - 1:200, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
| Rabbit | |
| 91 kDa | |
| 0.1 mg | |
| GPCR, Neuroendocrinology, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction | |
| 4915 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction