missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkB Antibody (8D2E8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £469.00
Specifications
| Antigen | TrkB |
|---|---|
| Clone | 8D2E8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18280581
|
Novus Biologicals
NBP2-52523 |
0.1 mg |
£469.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669697
|
Novus Biologicals
NBP2-52523-0.025mg |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TrkB Monoclonal specifically detects TrkB in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.Specifications
| TrkB | |
| 1 mg/mL | |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| GPCR, Neuroendocrinology, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 4915 | |
| Purified recombinant fragment of human NTRK2 (AA: 207-339) expressed in E. Coli. | |
| Primary | |
| 92 kDa |
| 8D2E8 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
| NTRK2 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title